Order Entry
Switzerland
ContactUsLinkComponent
D-(+)-Xylose, GPR RECTAPUR®
D-(+)-Xylose, GPR RECTAPUR®
Catalog # 29013.237
Supplier:  VWR Chemicals
CAS Number:  
undefined
D-(+)-Xylose, GPR RECTAPUR®
Catalog # 29013.237
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Melting point:
    145 - 150 °C
  • Specific optical rotation (10 %; water):
    18 - 20 °
  • Ignition residue (SO4):
    Max. 0.2 %
  • Loss on drying (100°C):
    Max. 0.2 %

Specification Test Results

About this item

 :
 :150.13 g/mol
Merck Index:13,10142
Boiling Pt:331 °C (1013 hPa)
Melting Pt:154 °C
Density:1.525 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-400-7
CAS Number:58-86-6
Formula:C₅H₁₀O₅
MDL Number:MFCD00151475