Order Entry
Switzerland
ContactUsLinkComponent
D-(+)-Glucose, anhydrous, GPR RECTAPUR®
D-(+)-Glucose, anhydrous, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
D-(+)-Glucose, anhydrous, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Spec. opt. rotation (calc. on anhydrous):
    52.5 - 53.3 °
  • Loss on drying (100°C):
    Max. 1 %
  • Ignition residue (SO4):
    Max. 0.1 %
  • Cl (Chloride):
    Max. 100 ppm

Specification Test Results

Frequently Bought Together