Order Entry
Switzerland
Orders LinkContactUsLinkComponent
D-(+)-Galactose 99+%

D-(+)-Galactose 99+%

D-(+)-Galactose 99+%
Certificates
D-(+)-Galactose 99+%

5 Options Available Below

Product Details & Documents
Synonyms:  D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
  • CAS Number:59-23-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00151230
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04356
  • Boiling Pt:527 °C (1013 hPa)
  • Melting Pt:165…168 °C
  • Density:1,5 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-416-4

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.3 % (K.F.)
  • Sulfated ash:
    ≤0.1 % (on dry substance)
  • Solubility:
    (10 % in water) Clear colorless
  • HPLC:
    ≥99.0 % (ELSD)
  • Specific optical rotation:
    +78° to +81.5° (20°C, 589 nm) (c=10, water, NH3) (after 30 minutes, on dry substance)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 75.00
    SFr. 75.00
    1 * 100 g
       SFr. 75.00

  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 260.00
    SFr. 260.00
    1 * 500 g
       SFr. 260.00

  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - SFr. 506.00
    SFr. 506.00
    1 * 1 kg
       SFr. 506.00

  • Plastic drum
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - SFr. 1’790.00
    SFr. 1’790.00
    1 * 5 kg
       SFr. 1’790.00

  • Fibre drum
    25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg - SFr. 5’990.00
    SFr. 5’990.00
    1 * 25 kg
       SFr. 5’990.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.