Deuterium oxide (99.9% D)
Catalog # CAMBDLM-4-1000
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:20 g/mol
Merck Index:12,02984
Boiling Pt:101.3 °C (1013 hPa)
Melting Pt:3.8 °C
Density:1.1 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:232-148-9
CAS Number:7789-20-0
Formula:D₂O
MDL Number:MFCD00044636