Dimethyl sulphoxide-[D₆] (99.9% D) +0.05% V/V TMS for NMR spectroscopy
Catalog # CAMBDLM-10TB-10
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:84.09 g/mol
Boiling Pt:189 °C (5 mmHg)
Melting Pt:20.2 °C
Density:1.19 g/cm³ (20 °C)
Flash Pt:82 °C (closed cup)
Storage Temperature:Ambient
EINECS:218-617-0
CAS Number:2206-27-1
Formula:D₃CS(O)CD₃
MDL Number:MFCD00002090