Order Entry
Switzerland
ContactUsLinkComponent
Chloroform-[D₁] (99.8% D) +0.05% V/V TMS for NMR spectroscopy
Health
Irritant
Chloroform-[D₁] (99.8% D) +0.05% V/V TMS for NMR spectroscopy
Catalog # CAMBDLM-7TB-100
CAS Number:  
Health
Irritant
Chloroform-[D₁] (99.8% D) +0.05% V/V TMS for NMR spectroscopy
Catalog # CAMBDLM-7TB-100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :120.38 g/mol
Boiling Pt:60.8 °C (1013 hPa)
Melting Pt:–64.1 °C
Density:1.5 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:212-742-4
UN:1888
ADR:6.1,III
 :GHS08
 :GHS07
CAS Number:865-49-6
Formula:CDCl₃
MDL Number:MFCD00000827