Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Beeswax, white, TECHNICAL

Plastic bottle, 500 g
Catalog # 22841.265
Supplier:   VWR Chemicals
Beeswax, white, TECHNICAL
Catalog # 22841.265
Supplier:  VWR Chemicals
CAS Number:  8012-89-3
Beeswax, white, TECHNICAL
Beeswax, white, TECHNICAL

Plastic bottle, 500 g

CAS Number:  8012-89-3
Price
SFr. 127.00
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Wax, bees
  • CAS Number:8012-89-3
  • MDL Number:MFCD00132754
  • Merck Index:12,01049
  • Melting Pt:∼64 °C
  • Density:0.95 g/cm³ (20 °C)
  • EINECS:232-383-7
Specifications

Specification Test Results

  • Melting point:
    61 - 65 °C

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.