



Petroleum spirit, 40…60 °C ≥75%, Puriss., meets analytical specification of DAB, low boiling point hydrogen treated naphtha, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849
Boiling Pt:40…60 °C (1.013 hPa)
Density:0.65 g/cm³ (20 °C)
Flash Pt:–30 °C
Storage Temperature:Ambient
EINECS:265-151-9
UN:3295
ADR:3,II
:GHS08
:GHS07
:GHS02
:GHS09
Specification Test Results
- Boiling range 40-60 °C:Min. 75 %
- Density (D 20/20):0.642 - 0.656
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.01 %
- Benzene (GC):Max. 0.001 %
- S-compounds (as S):Max. 0.005 %
- Appearance:complying
- Acidity or alkalinity:complying
- n-Hexane (GC):Max. 2 %
- Reaction against H₂SO₄:complying
- Tetraethyl lead:complying
- S-compounds, reducing matters:complying
Specification Test Results
Frequently Bought Together