Order Entry
Switzerland
ContactUsLinkComponent
 
Molecular sieve A5 (0.5 nm, 5 Å), powder < 50 µm
Catalog # ACRO214815000
CAS Number:  
undefined
Molecular sieve A5 (0.5 nm, 5 Å), powder < 50 µm
Catalog # ACRO214815000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • pH:
    10 to 10.8 (10 % suspension in water)
  • Residual water:
    ≤1.5 % (550°C, 2 h)
  • Water adsorption:
    ≥23 % (50%rH, 20°C, 24h)

Specification Test Results

About this item

 :
CAS Number:69912-79-4
 :1318-02-1