Order Entry
Switzerland
ContactUsLinkComponent
Molecular sieve A4 (0.4 nm, 4 Å), powder -325 mesh, activated, Fluka™
Molecular sieve A4 (0.4 nm, 4 Å), powder -325 mesh, activated, Fluka™
  688363-2.5KG
 :  
Molecular sieve A4 (0.4 nm, 4 Å), powder -325 mesh, activated, Fluka™
  688363-2.5KG
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg
  • Seq:
    4076

 

 

  • Form:
    powder
  • Sieve analysis < 0.045 mm:
    Min. 95 %

 

 

 :
 :400 °C
 :2.0 g/cm³
 :Ambient
 :70955-01-0
 :1318-02-1
 :MFCD00147627