Molecular sieve A4 (0.4 nm, 4 Å), powder <50 µm
:70955-01-0
:1318-02-1
:MFCD00131613
:1,1 to 2.2 g/cm³ (20 °C)
:Ambient
- Appearance:White to beige Powder
- pH:9 to 12 (10 % suspension in water)
- Residual water:≤3 %
- Water adsorption:≥18 %
Frequently Bought Together