Order Entry
Switzerland
ContactUsLinkComponent
Molecular sieve A4 (0.4 nm, 4 Å) 8 - 12 mesh
Molecular sieve A4 (0.4 nm, 4 Å) 8 - 12 mesh
Certificates
undefined
Molecular sieve A4 (0.4 nm, 4 Å) 8 - 12 mesh
Certificates

Specification Test Results

  • Appearance:
    Beige Beads
  • Bulk density:
    680 to 760 g/l
  • Residual water:
    ≤1.5 % (550°C)
  • Water adsorption:
    ≥20 % (1bar,25°C)

Specification Test Results

Related Information

Frequently Bought Together