Order Entry
Switzerland
ContactUsLinkComponent
Molecular sieve A4 (0.4 nm, 4 Å) 4 --> 8 mesh
Molecular sieve A4 (0.4 nm, 4 Å) 4 --> 8 mesh
Certificates
undefined
Molecular sieve A4 (0.4 nm, 4 Å) 4 --> 8 mesh

Specification Test Results

  • Appearance:
    Beige Beads
  • Bulk density:
    660 to 760 g/l
  • Residual water:
    ≤1.5 %
  • Water adsorption:
    ≥20 %

Specification Test Results

Related Information

Frequently Bought Together