Paraffin, liquid, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 7160.1000
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- Color (APHA):max. 20
- Identification (IR):passes test
Specification Test Results
About this item
Synonyms: Wax Paraffin, Paraffin wax
:
Merck Index:13,07093
Boiling Pt:300…500 °C (1013 hPa)
Density:0.86 g/cm³ (20 °C)
Flash Pt:215 °C (closed cup)
EINECS:232-384-2
CAS Number:8012-95-1
:8002-74-2