Molecular sieve 13X (1.0 nm, 10 Å)
Catalog # WAKO131-07085
Supplier: Wako
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
CAS Number:63231-69-6
:1318-02-1
MDL Number:MFCD00164506