Molecular sieve 13X (1.0 nm, 10 Å), beads 1.6-2.5 mm (0.063-0.098 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:63231-69-6
:1318-02-1
MDL Number:MFCD00164506
Storage Temperature:Ambient
Frequently Bought Together