Order Entry
Switzerland
ContactUsLinkComponent
Decahydronaphthalene (mixture of cis and trans isomers) 98%, Reagent Grade
Corrosive
Health
Toxic
Flammable
Environment
Decahydronaphthalene (mixture of cis and trans isomers) 98%, Reagent Grade
Certificates
undefined
Corrosive
Health
Toxic
Flammable
Environment
Decahydronaphthalene (mixture of cis and trans isomers) 98%, Reagent Grade

Specification Test Results

  • Vapor Pressure:
    741 mmHg ( 188 °C)
  • Identity (IR):
    complying
  • Assay on cis/trans-isomers (GC):
    Min. 97.5 %
  • Explosion Limit:
    0.7-4.9 %, 100 °F
  • Refractive Index:
    n20/D 1.474 (lit.)
  • Vapor Density:
    4.76 (vs air)

Specification Test Results

Frequently Bought Together