



Decahydronaphthalene (mixture of cis and trans isomers) ≥99%, TECHNICAL
Catalog # 23269.297
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay (calculated as mixture of isomers):Min. 99 %
- Refractive index (20°C):1.473 - 1.475
Specification Test Results
About this item
:
:138.25 g/mol
Merck Index:13,02866
Boiling Pt:189…191 °C (1013 hPa)
Melting Pt:∼–40 °C
Density:0.8696 g/cm³ (20 °C)
Flash Pt:57 °C
Storage Temperature:Ambient
EINECS:202-046-9
UN:1147
ADR:3,III
REACH:01-2119565127-37
:GHS02
:GHS07
:GHS05
:GHS09
CAS Number:91-17-8
Formula:C₁₀H₁₈
MDL Number:MFCD00004130