Order Entry
Switzerland
ContactUsLinkComponent
Decahydronaphthalene (mixture of cis and trans isomers) for synthesis, Sigma-Aldrich®
Flammable
Toxic
Corrosive
Health
Environment
Decahydronaphthalene (mixture of cis and trans isomers) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Flammable
Toxic
Corrosive
Health
Environment
Decahydronaphthalene (mixture of cis and trans isomers) for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    0.89 mg/l (25 °C) (experimental)
  • Melting Point:
    -40 °C
  • Molar Mass:
    138.25 g/mol
  • Boiling Point:
    189 - 191 °C (1013 hPa)
  • Vapor Pressure:
    1.3 hPa (20 °C)
  • Flash Point:
    57 °C
  • Refractive Index:
    1.4742 (20 °C, 589 nm)
  • Explosion Limit:
    0.7 - 4.9 %(V)
  • Density:
    0.88 g/cm3 (20 °C)
  • Ignition Point:
    255 °C
  • Viscosity kinematic:
    2.2 mm2/s (25 °C)
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specification Test Results

Related Information

Frequently Bought Together