




Decahydronaphthalene (mixture of cis and trans isomers) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:91-17-8
:
Formula:C₁₀H₁₈
MDL Number:MFCD00004130
:138.25 g/mol
Merck Index:14,2846
Boiling Pt:189…191 °C
Melting Pt:< –45 °C
Density:0.88
Flash Pt:57 °C (134 °F)
EINECS:202-046-9
UN:1147
ADR:3,III
:GHS06
:GHS02
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together