Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
 
 
Decahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture 98%
 
undefined
 
 
 
 
 
Decahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture 98%

 

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4730 to 1.4760 (20°C, 589 nm)
  • Water:
    ≤0.1 %
  • GC:
    ≥97.5 %
  • Color scale:
    ≤10 APHA

 

 

Frequently Bought Together