tert-Butyl-4-methoxyphenol (mixture of isomers)
EHERC10931220
:
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
:
:180.25 g/mol
:12,01582
:264 °C (1013 hPa)
:48…55 °C
:1.05 g/cm³ (20 °C)
:156 °C
:Ambient
:246-563-8
:25013-16-5
:C₁₁H₁₆O₂
:MFCD01779059