Platinum ≥99.997% (metals basis), foil, Premion®, Thickness: 0.5 mm (0.02 in)
Catalog # 12057.FF
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:1643
- Environmentally Preferable:
- Pk Info:25×25 mm
Specifications
About this item
Synonyms: Platinium
:
:195.08 g/mol
Merck Index:12,07689
Boiling Pt:3827 °C (1013 hPa)
Melting Pt:1769 °C
Density:21.447 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-116-1
CAS Number:7440-06-4
Formula:Pt
MDL Number:MFCD00011179