Platinum ≥99.9% (metals basis), wire, hard, Ø 0.127 mm (0.005 in)
: Platinium
:7440-06-4
:
:Pt
:MFCD00011179
:195.08 g/mol
:12,07689
:3827 °C (1013 hPa)
:1769 °C
:21.447 g/cm³ (20 °C)
:Ambient
:231-116-1
- Environmentally Preferable:
Frequently Bought Together