Order Entry
Switzerland
ContactUsLinkComponent
VWR®, Cryoboxes, PP, with Dividers
VWR®, Cryoboxes, PP, with Dividers
Catalog # 479-1349
Supplier:  VWR Collection
undefined
VWR®, Cryoboxes, PP, with Dividers
Catalog # 479-1349
Supplier:  VWR Collection

Specifications

  • Compartments:
    81 (9×9)
  • For tube Ø:
    12 mm
  • For tubes:
    1,5/2,0 ml
  • Colour:
    White
  • Pk:
    10

Specifications

About this item

Made from corrugated PP.

  • Significantly reduces the chance of mold
  • Dividers included with the boxes
  • Store down to temperatures of -80 °C
  • Can be used in mechanical freezers and vessels down to -196 ºC ( Vapour Phase only)

This easy self assemble cryo storage box is flat packed in design and offers compact size.

Since corrugated PP is not susceptible to moisture, durability is higher than card boxes.