Order Entry
Switzerland
ContactUsLinkComponent
Cobalt ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Health
Cobalt ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 40835.KS
CAS Number:  
Health
Cobalt ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 40835.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7440-48-4
 :
Formula:Co
MDL Number:MFCD00010935
 :58.93 g/mol
Merck Index:12,02488
Boiling Pt:2870 °C (1013 hPa)
Melting Pt:1495 °C
Density:8.9 g/cm³ (20 °C)
EINECS:231-158-0
 :GHS08
Storage Temperature:Ambient