Order Entry
Switzerland
ContactUsLinkComponent
Manganese, powder, Sigma-Aldrich®
Flammable
Manganese, powder, Sigma-Aldrich®
Catalog # 1.12237.0250
Supplier:  Merck
CAS Number:  
Flammable
Manganese, powder, Sigma-Aldrich®
Catalog # 1.12237.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    1245 °C
  • Molar Mass:
    54.94 g/mol
  • Boiling Point:
    2097 °C
  • Density:
    7.2 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :54.94 g/mol
Merck Index:12,05762
Boiling Pt:2097 °C (1013 hPa)
Melting Pt:1260 °C
Density:7.2 g/cm³ (20 °C)
EINECS:231-105-1
UN:3089
ADR:4.1,
 :M-Clarity™ quality level = MQ200
 :GHS02
CAS Number:7439-96-5
Formula:Mn
MDL Number:MFCD00011111