
Manganese 99+%, powder -40 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-96-5
:
Formula:Mn
MDL Number:MFCD00011111
:54.94 g/mol
Merck Index:12,05762
Boiling Pt:1900 °C
Melting Pt:1260 °C
Density:7,2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-105-1
UN:3089
ADR:4.1,III
:GHS02
Specification Test Results
- Appearance:Grey-brown to brown-black Powder
- Assay:≥99.0 % (Mn)
Specification Test Results
Frequently Bought Together