Order Entry
Switzerland
ContactUsLinkComponent
Manganese ≥99.95% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
Manganese ≥99.95% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 40864.KS
CAS Number:  
Manganese ≥99.95% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 40864.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7439-96-5
 :
Formula:Mn
MDL Number:MFCD00011111
 :54.94 g/mol
Storage Temperature:Ambient
EINECS:231-105-1