Order Entry
Switzerland
ContactUsLinkComponent
Molybdenum ≥99.95% (metals basis), wire, annealed, Ø 0.375 mm (0.015 in)
Molybdenum ≥99.95% (metals basis), wire, annealed, Ø 0.375 mm (0.015 in)
Catalog # 43996.8V
CAS Number:  
Molybdenum ≥99.95% (metals basis), wire, annealed, Ø 0.375 mm (0.015 in)
Catalog # 43996.8V
CAS Number:  

Specifications

  • Pk:
    60 m
  • Seq:
    8426

Specifications

About this item

CAS Number:7439-98-7
 :
Formula:Mo
MDL Number:MFCD00003465
Merck Index:12,06317
Boiling Pt:3617 °C (1013 hPa)
Melting Pt:2622 °C
Density:10.28 g/cm³ (20 °C)
 :95.94 g/mol
Storage Temperature:Ambient
EINECS:231-107-2