Order Entry
Switzerland
ContactUsLinkComponent
Molybdenum ≥99.95% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Molybdenum ≥99.95% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Catalog # 42358.22
CAS Number:  
Molybdenum ≥99.95% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Catalog # 42358.22
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Merck Index:12,06317
Boiling Pt:3617 °C (1013 hPa)
Melting Pt:2622 °C
Density:10.28 g/cm³ (20 °C)
 :95.94 g/mol
Storage Temperature:Ambient
EINECS:231-107-2
CAS Number:7439-98-7
Formula:Mo
MDL Number:MFCD00003465