
Molybdenum ≥99.95% (metals basis), powder APS 3-7 µm
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-98-7
:
Formula:Mo
MDL Number:MFCD00003465
Boiling Pt:5560 °C (1013 hPa)
Melting Pt:2622 °C
Density:10.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-107-2
UN:3089
ADR:4.1,III
:95.94 g/mol
:GHS02
Related Information
Frequently Bought Together