

Vanadium ≥99.5% (metals basis), powder -325 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-62-2
:
Formula:V
MDL Number:MFCD00011453
:50.94 g/mol
Boiling Pt:3000 °C
Melting Pt:1900 °C
Density:6.11
Storage Temperature:Ambient
EINECS:231-171-1
UN:3089
ADR:4.1,II
:GHS02
:GHS07
Related Information
Frequently Bought Together