Order Entry
Switzerland
ContactUsLinkComponent
Tantalum ≥99.95% (metals basis excluding Nb), sputtering target 76.2 mm (3.0 in) Ø x 3.18 mm (0.125 in) thick,
Tantalum ≥99.95% (metals basis excluding Nb), sputtering target 76.2 mm (3.0 in) Ø x 3.18 mm (0.125 in) thick,
Catalog # 40896.KS
CAS Number:  
Tantalum ≥99.95% (metals basis excluding Nb), sputtering target 76.2 mm (3.0 in) Ø x 3.18 mm (0.125 in) thick,
Catalog # 40896.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :180.95 g/mol
Merck Index:12,09223
Boiling Pt:5427 °C (1013 hPa)
Melting Pt:2996 °C
Density:16.69 g/cm³ (20 °C)
EINECS:231-135-5
CAS Number:7440-25-7
Formula:Ta
MDL Number:MFCD00011252