Order Entry
Switzerland
ContactUsLinkComponent
Sodium hydroxide solution 40% (w/w)
Corrosive
Sodium hydroxide solution 40% (w/w)
Supplier:  PanReac AppliChem
Certificates
Corrosive
Sodium hydroxide solution 40% (w/w)
Supplier:  PanReac AppliChem
Certificates

Specification Test Results

  • Minimum Assay (Acidim.):
    40%
  • Nitrogen Compounds (as N):
    Max. 0.002%
  • Carbonate (as Na₂CO₃):
    Max. 1.0%

Specification Test Results

Related Information

Frequently Bought Together