Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Sulphur

Irritant
Marketsource
Supplier:   Roth Carl
Sulphur
Supplier:  Roth Carl
Marketsource
Certificates

3 Options Available Below

Product Details & Documents
  • CAS Number:7704-34-9
  • Formula:S
  • MDL Number:MFCD00085316
  • Molecular Weight:32.07 g/mol
  • Merck Index:13,09059
  • Boiling Pt:445 °C (1013 hPa)
  • Melting Pt:113…119 °C
  • Density:2.36 g/cm³ (20 °C)
  • Flash Pt:168 °C
  • Storage Temperature:Ambient
  • EINECS:231-722-6
  • UN:1350
  • ADR:4.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - SFr. 25.80
    SFr. 25.80
    1 * 1 kg
       SFr. 25.80

    Specifications:
    Marketsource
    Marketsource
  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - SFr. 83.40
    SFr. 83.40
    1 * 5 kg
       SFr. 83.40

    Specifications:
    Marketsource
    Marketsource
  • 25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg - SFr. 238.80
    SFr. 238.80
    1 * 25 kg
       SFr. 238.80

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.