Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Sulphur ≥99%, flower, TECHNICAL

Sulphur ≥99%, flower, TECHNICAL

Plastic bottle for solids, 1 kg
Catalog # 28256.296
Irritant
Supplier:   VWR Chemicals
Sulphur ≥99%, flower, TECHNICAL
Catalog # 28256.296
Supplier:  VWR Chemicals
CAS Number:  7704-34-9
Sulphur ≥99%, flower, TECHNICAL
Sulphur ≥99%, flower, TECHNICAL

Plastic bottle for solids, 1 kg

CAS Number:  7704-34-9
Price
SFr. 84.20
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:7704-34-9
  • Formula:S
  • MDL Number:MFCD00085316
  • Molecular Weight:32.07 g/mol
  • Merck Index:13,09059
  • Boiling Pt:445 °C (1013 hPa)
  • Melting Pt:113…119 °C
  • Density:2.36 g/cm³ (20 °C)
  • Flash Pt:168 °C
  • Storage Temperature:Ambient
  • EINECS:231-722-6
  • UN:1350
  • ADR:4.1,III
Specifications

Specification Test Results

  • Assay:
    Min. 99 %

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg

Specifications