Serological pipettes, Shorty, KIMBLE®
Shorty pipettes are made from glass and are ideal for use in tissue culture work and other procedures performed under a fume hood or in other confined areas.
- 'Reverse' and 'negative' graduations increase the versatility of the pipette
- Calibrated 'to deliver' with blow-out after free flow of liquid has ended
- Multi-pack bag: One or several pipettes can be removed without contaminating the entire pack
- Non pyrogenic, sterile, disposable
: Individually wrapped in paper/plastic or packed in a multi-pack bag.
Frequently Bought Together