Aktivkohle (pflanzliche Basis), Pulver, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:64365-11-3
Formula:C
MDL Number:MFCD00133992
:12.01 g/mol
Merck Index:13,01818
Melting Pt:3550 °C
Density:2 g/cm³ (20 °C)
EINECS:231-153-3
UN:1362
ADR:4.2,III
REACH:01-2119488894-16
Specification Test Results
- Mean particle size:Max. 80 µm
- Ignition residue:Max. 6 %
- Water:Max. 10 %
Specification Test Results
Related Information
Certificates
Search by Lot Number22637_CHARCOAL VEGETAL ACTIVATED POWDER_BSE_REV0
BSE/TSE-Zertifikate
SDS_DE_CH_VWRC22637.pdf sds_VWRC22637
Sicherheitsdatenblatt (SDS)
spec_VWRC22637.293 spec_VWRC22637.293 spec_VWRC22637.415
Produktspezifikation
VWR® for Drying and Filtration
Produktliteratur
Frequently Bought Together