
Indium ≥99.99% (metals basis), powder -100 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-74-6
:
Formula:In
MDL Number:MFCD00134048
:114.82 g/mol
Boiling Pt:2072 °C
Melting Pt:156.6 °C
Density:7.31
Storage Temperature:Ambient
EINECS:231-180-0
UN:3089
ADR:4.1,III
:GHS02
Related Information
Frequently Bought Together