
Aluminium, powder for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7429-90-5
:
Formula:Al
MDL Number:MFCD00134029
:26.98 g/mol
Density:2.7 g/cm³ (20 °C)
EINECS:231-072-3
UN:1396
ADR:4.3,II
:M-Clarity™ quality level = MQ200
:GHS02
Specification Test Results
- Solubility:(20 °C) (reaction)
- Molar Mass:26.98 g/mol
- Hazard Class:4.3
- Hazard Class Description:Dangerous When Wet Material
Specification Test Results
Related Information
Frequently Bought Together