Order Entry
Switzerland
ContactUsLinkComponent
Aluminium ≥99.9999% (metals basis), wire, Puratronic®, annealed, Ø 0.5 mm (0.02 in)
Aluminium ≥99.9999% (metals basis), wire, Puratronic®, annealed, Ø 0.5 mm (0.02 in)
Catalog # 44333.G6
CAS Number:  
Aluminium ≥99.9999% (metals basis), wire, Puratronic®, annealed, Ø 0.5 mm (0.02 in)
Catalog # 44333.G6
CAS Number:  

Specifications

  • Pk:
    10 m
  • Seq:
    8390

Specifications

About this item

CAS Number:7429-90-5
 :
Formula:Al
MDL Number:MFCD00134029
 :26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (1013 hPa)
Melting Pt:660 °C
Density:2.7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-072-3