Auftragserfassung
Switzerland
ContactUsLinkComponent
Flammable
Magnesium ≥98,5%, Pulver, Ø ~ 0.06-0.3 mm, Supelco®
Catalog # 1.05815.1000
Supplier:  Merck
CAS Number:  
Flammable
Magnesium ≥98,5%, Pulver, Ø ~ 0.06-0.3 mm, Supelco®
Catalog # 1.05815.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Packung:
    Metalldose
  • VE:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay (complexometric):
    ≥ 98.5 %
  • Substances insoluble in hydrochloric acid:
    ≤ 0.05 %
  • Fe (Iron):
    ≤ 500 ppm

Specification Test Results

About this item

 :
 :24.3 g/mol
Boiling Pt:1107 °C (1013 hPa)
Melting Pt:651 °C
Density:1.74 g/cm³ (25 °C)
Flash Pt:500 °C
Storage Temperature:Raumtemperatur
EINECS:231-104-6
UN:1418
ADR:4.3 (4.2),II
 :M-Clarity™ quality level = MQ200
 :GHS02
CAS Number:7439-95-4
Formula:Mg
MDL Number:MFCD00085308