ContactUsLinkComponent
Sodium hydroxide 0.5 mol/l (0.5 N) in water volumetric solution
Sodium hydroxide 0.5 mol/l (0.5 N) in water volumetric solution
  • Titer at 20 °C:
    0.999 - 1.001
  • Uncertainty:
    See Certificate
  • Traceability:
    NIST
  • pH:
    13.5
Frequently Bought Together