



Trimethyl[3-(trimethoxysilyl)propyl]ammonium chloride 50% in methanol
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:35141-36-7
:
Formula:C₉H₂₄ClNO₃Si
MDL Number:MFCD00054225
:257.83 g/mol
Density:0.930
Flash Pt:11 °C (52 °F)
Storage Temperature:Ambient
EINECS:252-393-5
UN:1230
ADR:3,II
:GHS02
:GHS06
:GHS08
:GHS07
Specification Test Results
- Assay (Non-Aqueous Acid-Base Titration):48.0 - 52.0%
- Refractive Index:1.3970
Specification Test Results
Frequently Bought Together