Orders LinkContactUsLinkComponent
Waste bins, Hospisafe
Waste bins, Hospisafe
Catalog #
Waste bins, Hospisafe
Catalog #
Supplier:  
Waste bins, Hospisafe
Waste bins, Hospisafe
Product Details & Documents
The containers are made of pure PP, when burned no pollutants are released.
  • Leakproof and gastight after closing
  • Very good chemical resistance
Certifications: UN approved and conforms with NFX 30511, ADR and NFX 30505.