Organic analytical standard, estragole, Supelco®
34098-1ML
: Merck
:
- VE:1 mL
- Normen:Reference standard
- CAS Nr.:140-67-0
- Bezeichnung:Estragole, ≥98%, analytical standard
- M-Clarity Qualitätsstufe:MQ100
- Formel:C₁₀H₁₂O
- Bezeichnung:Estragole, analytical standard
Synonym: 4-allylanisole, chavicol methyl ether, estragole.
- Refractive index: n20/D 1,521 (lit.)
- Boiling point: 215 – 216 °C (lit.)
- Density: 0,965 g/ml at 25 °C (lit.)
: Shelf life: limited shelf life, expiry date on the label