Orders LinkContactUsLinkComponent
Automatic Burettes, Pellet Pattern, Class AS
Automatic Burettes, Pellet Pattern, Class AS
Supplier:   DWK Life Sciences
Automatic Burettes, Pellet Pattern, Class AS
Supplier:  DWK Life Sciences
Certificates
Automatic Burettes, Pellet Pattern, Class AS
Automatic Burettes, Pellet Pattern, Class AS
6 Options Available Below
Product Details & Documents
Burette, automatic, Pellet, DURAN® borosilicate glass, with PTFE key, class AS.
  • 30 seconds waiting time
  • Schellbach stripe and main graduations as circular divisions
  • Supplied with reservoir bottle and blow ball
Certifications: DIN EN ISO 385
Documents
Product Family Options
Without reservoir bottle and blowball
Product Information
  • Capacity
    Division
    Length
    Tolerance
    Availability
    Price
  • 10 ml
    0,02 ml
    912 mm
    0,02 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 383.00
    SFr. 383.00
    1 * 1 ST
       SFr. 383.00
  • 25 ml
    0,05 ml
    912 mm
    0,03 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 403.00
    SFr. 403.00
    1 * 1 ST
       SFr. 403.00
  • 50 ml
    0,1 ml
    912 mm
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 403.00
    SFr. 403.00
    1 * 1 ST
       SFr. 403.00
Product Information
  • Capacity
    Division
    Length
    Tolerance
    Availability
    Price
  • 10 ml
    0,02 ml
    930 mm
    0,02 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 459.00
    SFr. 459.00
    1 * 1 ST
       SFr. 459.00
  • 25 ml
    0,05 ml
    930 mm
    0,03 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 459.00
    SFr. 459.00
    1 * 1 ST
       SFr. 459.00
  • 50 ml
    0,1 ml
    930 mm
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 459.00
    SFr. 459.00
    1 * 1 ST
       SFr. 459.00
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.