Order Entry
Switzerland
ContactUsLinkComponent
Cup, large with handle
Cup, large with handle
Catalog # LAMA2147
Supplier:  LAMAPLAST
undefined
Cup, large with handle
Catalog # LAMA2147
Supplier:  LAMAPLAST

Specifications

  • Capacity:
    3000 ml
  • Description:
    Cup, length 115 mm
  • Pk:
    1

Specifications

About this item

Made of HDPE, white.

  • With handle
  • Easy to clean

For food applications.