Order Entry
Switzerland
ContactUsLinkComponent
Reference standard for food and environment analysis, disperse orange 61, Dr Ehrenstorfer
Reference standard for food and environment analysis, disperse orange 61, Dr Ehrenstorfer
Catalog # EHERC12972135
Supplier:  EHRENSTORFER
CAS Number:  
Reference standard for food and environment analysis, disperse orange 61, Dr Ehrenstorfer
Catalog # EHERC12972135
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    100 mg
  • CAS No.:
    55281-26-0
  • Description:
    Disperse orange 61
  • Formula:
    C₁₇H₁₅Br₂N₅O₂
  • Description:
    Disperse orange 61

Specifications

About this item